Syringe filters, prefilter, non-sterile, GD/X™, Whatman™
Toimittaja: Whatman products (Cytiva)
6870-1302 6870-1304 6870-2502 6870-2504 6871-1302 6871-1304 6871-2502 6871-2504 6872-1302 6872-1304 6872-2502 6872-2504 6873-1304 6873-2502 6873-2504 6874-1302 6874-1304 6874-2502 6874-2504 6875-1302 6875-1304 6875-2502 6875-2504 6876-1302 6876-2502 6876-2504 6878-1304 6880-1302 6880-2502 6880-2504 6881-2504 6882-1316 6882-2516 6884-1310 6884-2510 6886-2512 6888-1327 6888-2527 6890-1307 6890-2507 6892-2515 6894-1304 6894-2504 6895-2504 6870-2550 6879-2504 6883-2516 6891-2507 6905-2502 6905-2504 6882-2504 6883-2504 6887-2502 6871-2550
514-8039EA 729 EUR
514-8039 514-8040 514-8041 514-8042 516-6312 516-6313 516-6314 516-6315 514-8043 514-8044 514-8045 514-8046 516-6317 516-6318 516-6319 514-8047 514-8048 514-8049 514-8050 516-6320 516-6321 516-6322 514-8051 514-8052 514-8054 514-8055 514-8057 514-8060 514-8062 514-8063 516-6324 516-1373 514-0528 514-0529 514-0530 514-0531 514-0532 514-0533 514-0534 514-0535 516-1372 514-0536 514-8064 516-6309 514-0680 514-0290 516-0990 516-0992 514-0291 514-0292 511-0161 516-0414 511-0162 WHAT6871-2550
Syringe filters, prefilter, non-sterile, GD/X™, Whatman™
Suodattimet Ruiskusuodattimet
Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.
  • Recyclable after use
Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- CelluloseMore Product Information