PES centrifugal concentrator, Vivaspin® 100
Toimittaja: Sartorius
VC1051 VC1072 VC1052 VC1042 VC1041 VC1032 VC1031 VC1022 VC1021 VC1012 VC1061 VC1001 VC1011 VC1002
516-0138EA 124 EUR
516-0138 516-0133 516-0137 516-0139 516-0140 516-0141 516-0142 516-0143 516-0144 516-0145 516-0136 516-0148 516-0146 516-0147
PES centrifugal concentrator, Vivaspin® 100
Suodattimet Sentrifugisuodattimet
Vivaspin® 100 PES centrifugal concentrators are versatile ultrafiltration units for centrifugation-based sample concentration as well as pressure filtration applications.
  • Easy to assemble and handle
  • High flux PES membranes
  • High target molecule recoveriesLargest capacity centrifugal device; for samples up to 90 ml
  • Total process flexibility
  • Constructed of PC body and filtrate vessel, PP concentrator cap
Popular applications for Vivaspin® centrifugal concentrators:
- Concentration | diafiltration of proteins, enzymes, nucleic acids, monoclonal antibodies,More Product Information