Glass microfiber filters with organic binder, grade MN 85/90
Toimittaja: MACHEREY-NAGEL
405005 405009 405024 405011 405012 405018 4050025 4050037 4050045 405007 405015 405027
MANA405005EA 53.1 EUR
MANA405005 515-9795 MANA405024 515-9796 515-9797 515-9798 MANA4050025 MANA4050037 MANA4050045 MANA405007 MANA405015 MANA405027
Glass microfiber filters with organic binder, grade MN 85/90
Suodattimet Lasikuitusuodattimet
The MN 85/90 grade is the most widely used, general purpose glass fiber filter with very good mechanical stability. The filter contains an organic binder.
  • High mechanical stability
  • General purpose filter
  • Stable up to 200 °C