Grade 2294 filter papers for technical use, Whatman™
Toimittaja: Whatman products (Cytiva)
10342810 10342860 10342862
515-8062EA 236 EUR
515-8062 WHAT10342860 WHAT10342862
Grade 2294 filter papers for technical use, Whatman™
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
A very thick filter card with high wet strength. Offers very high flow rate and retains medium to coarse particles.
  • With central hole
The applications range from the filtration of beverages to the purification of electroplating baths.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application.