Qualitative filter papers, standard grades, grade 604 ½, Whatman™
Toimittaja: Whatman products (Cytiva)
10312744 10312745 10312747 10312751 10312753
515-5004EA 77.7 EUR
515-5004 515-5005 515-5006 515-5007 515-5009
Qualitative filter papers, standard grades, grade 604 ½, Whatman™
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
Grade 604½ Qualitative Filter Papers for coarse precipitates. Maintains flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
  • Increased total loading capacity as more filter area is available
Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.