Qualitative filter papers, technical, smooth, grade 3 w
Toimittaja: Sartorius
FT2-308-580580 FT-4-308-320 FT-4-308-270 FT-4-308-240 FT-4-308-185 FT-4-308-150 FT-4-308-125 FT-4-308-110 FT-3-308-270 FT-3-308-320 FT-3-308-240 FT-3-308-185 FT-3-308-150 FT-3-308-125 FT-3-308-110 FT-3-308-090 FT-3-308-070 FT-3-308-055
515-1620EA 130 EUR
515-1620 SARTFT-4-308-320 SARTFT-4-308-270 SARTFT-4-308-240 SARTFT-4-308-185 SARTFT-4-308-150 SARTFT-4-308-125 SARTFT-4-308-110 SARTFT-3-308-270 SARTFT-3-308-320 SARTFT-3-308-240 SARTFT-3-308-185 SARTFT-3-308-150 SARTFT-3-308-125 SARTFT-3-308-110 SARTFT-3-308-090 SARTFT-3-308-070 SARTFT-3-308-055
Qualitative filter papers, technical, smooth, grade 3 w
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration. Used for boric acid solution; essences; hair tonics; eau de cologne; tinctures.