Suodatinsuppilo MicroFunnel™ Plus
Toimittaja: Cytiva (Formerly Pall Lab)
4807 4808 4809 4814 4823 4844 4807ME 4829ME
516-8964EA 577 EUR
516-8964 516-9890 516-9891 515-0052 515-0055 515-0157 516-1238 516-1239
Suodatinsuppilo MicroFunnel™ Plus
Suppilot Membraanisuodatinsuppilot
Ideal for monitoring purified water systems maintained at temperatures up to 90 °C (194 °F).
  • Multifunctional
  • Leakproof lid
  • Squeeze separation
  • Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0,2 and 0,45 µm Supor® (More Product Information