Ruiskusuodatin, GD/XP™
Toimittaja: Whatman products (Cytiva)
6992-2504 6994-2504 6970-2504 6972-2504 6974-2504 6971-2504 6973-2504 6993-2504 6995-2504
514-8068EA 686 EUR
514-8068 514-8069 514-8065 516-1319 514-8066 514-0293 516-5115 516-5117 516-5124
Ruiskusuodatin, GD/XP™
Suodattimet Ruiskusuodattimet
These syringe filters, with pigment-free PP housing, are ideal for use with samples that require inorganic ion analysis, as levels of ion extractables are minimised.
  • Extremely low protein binding
  • Contain a two-layer pre-filter stack comprising of 20 and 5 µm PP filters, with a choice of membranes
  • High loading volume for filtration of challenging samples
  • Flow should enter from the inlet
Because of their high purity and minimal ion extractables, GD/XP syringe filters are recommended for analysing ion content that would otherwise experience interferenceMore Product Information