Syringe filters, PTFE, Puradisc™, Whatman™
Toimittaja: Whatman products (Cytiva)
6775-1304 6783-0402 6783-0404 6783-1302 6783-1304 6784-0402 6784-0404 6784-1301 6784-1302 6784-1304 6784-1310 6784-2501 6784-2502 6784-2504 6784-2510 6785-2502 6785-2504 10462300 6784-1350 6798-2504 6766-1302 6766-1304 6798-2501 6798-2502 6798-2510
514-0498EA 225 EUR
514-0498 516-1153 516-1155 514-0512 514-0513 516-1152 516-1154 516-1251 516-1252 516-1254 516-1257 514-8023 514-8024 514-8025 514-8026 514-8027 514-8028 514-1101 516-0435 514-0246 516-0920 516-0921 514-0244 514-0245 514-0247
Syringe filters, PTFE, Puradisc™, Whatman™
Suodattimet Ruiskusuodattimet
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample typesMore Product Information