Ruiskusuodatin, Acrodisc®, Supor®
Toimittaja: Cytiva (Formerly Pall Lab)
4187 4504 4506 4508 4509 4525 4602 4604 4611 4612 4614 4618 4650 4651 4652 4653 4654 4655 4656 4658 4659 4661 4662 4608 AP-4426 4668 AP-4425 AP-4799 4692
514-4102EA 190 EUR
514-4102 516-7654 516-7655 516-7656 516-7657 514-4119 514-4122 514-4123 514-4125 514-4126 514-4127 514-4128 514-4129 514-4130 514-4131 514-4132 514-4133 514-4134 514-4135 514-4136 516-7701 516-7703 516-9087 514-4124 514-0862 517-0003 514-0861 514-1105 PALL4692
Ruiskusuodatin, Acrodisc®, Supor®
Suodattimet Ruiskusuodattimet
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
  • Superior flow rates and higher throughputs than competitive devices
  • Low protein binding to minimize sample loss
  • Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
  • A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
  • Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
  • Reduces mycoplasma with the