Tube top vacuum filtration systems, Sartolab® RF 50
Toimittaja: Sartorius
180E01-2 180F01-2 180E01-E8 180F01-E8
514-1194EA 137 EUR
514-1194 514-1195 514-1459 514-1460
Tube top vacuum filtration systems, Sartolab® RF 50
Suodattimet Pullosuodattimet
Sartolab® RF 50 vacuum filtration units are single-use units for the filtration of sample volumes up to 50 ml.
They are available with either a 0,22 or 0,45 µm polyethersulfone membrane.
  • Highest flow rates with an asymmetric membrane and a large filtration surface
  • No loss of protein with a low protein affinity membrane
  • Low dead volume due to an optimized membrane support
  • Minimised risk of contamination with a complete ready-to-use unit (no additional transfer of liquid necessary for storage)
The design of the yellow adapter connecting the funnel to theMore Product Information