Acrodisc™ PSF Non-Sterile RC Syringe Filters
Toimittaja: Cytiva (Formerly Pall Lab)
AP-4144 AP-4145 AP-4146 AP-4147
514-1444EA 3030 EUR
514-1444 514-1445 514-1446 514-1447
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Suodattimet Ruiskusuodattimet
Acrodisc™ PSF high performance syringe filters can be used for general clarification or application specific uses like dissolution, HPLC, and IC. Engineered to work in most automated filter systems.
  • Regenerated cellulose (RC) membrane syringe filters (non-sterile) designed for automated filtration systems
  • Polypropylene housing ensures smooth transport from storage turntable to the filtration site
  • Regenerated cellulose-Borosilicate glass filter media
  • Use as a prefilter for complex samples
  • Enhance your HPLC column's life by using a prefilter
  • Optimised