Storage bottles, single-use, polystyrene, Sartolab®
Toimittaja: Sartorius
180-22-E 180-23-E 180-24-E 180-25-E
514-1294EA 38.9 EUR
514-1294 514-1295 514-1296 514-1297
Storage bottles, single-use, polystyrene, Sartolab®
Pullot Muovipullot
Single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
  • Low-profile design for easy gripping with one hand
  • Ergonomically designed cap to provide a rigid grip and one-hand handling of each bottle
  • The bottles with an indented bottom are stackable to save space in the refrigerator and in the freezer
  • Caps (45 mm) with airtight seal to minimise the risk of contamination
  • Graduation marks on the bottles ensure accuracy and easy reading
  • Designated writing field for clear labeling of samples
  • The materials used in the production