VWR®, Pullosuodattimet, PES
Toimittaja: VWR Collection
514-0328EA 124 EUR
514-0328 514-0329 514-0330 514-0331 514-0332 514-0333 514-0334 514-0335 514-0336 514-0337 514-0338 514-0339 514-0340 514-0341 514-0342 514-0343 514-0344 514-0345 514-0346 514-0347
VWR®, Pullosuodattimet, PES
Suodattimet Pullosuodattimet
PS funnel and collection reservoir. Systems for vacuum filtration of aqueous solutions including cell culture media, buffers or other biological fluids.
  • Each individual unit is lot numbered for traceability
  • Once filtration is complete, solution can be stored in the collection reservoir until needed
  • Non pyrogenic, non cytotoxic
PES membrane: Low protein binding and low extractables. Ideal for tissue culture applications.
0,2 µm: Ideal for sterilisation applications and media preparation.
0,45 µm: Ideal for buffer clarification.