Vacuum filtration units, GV 050, Whatman™
Toimittaja: Whatman products (Cytiva)
10442100
514-0115EA 838 EUR
514-0115
Vacuum filtration units, GV 050, Whatman™
Kolvit ja erlenmeyerit Imupullot
Cytiva offers a choice of Whatman holders for use with membrane filters.
  • Chemically resistant to most aqueous and organic solutions
  • Resistant to acids and caustic solutions
  • Autoclavable and can be sterilized in dry heat to 180°C
  • Can be used up to 200°C
Vaccum filtration devices made of borosilicate glass with silicone cover, suitable for analyses of residues, contamination tests and particulate analyses. Glass material. Silicone cap material. PTFE and silicone seals. Stainless steel, PTFE coated sieve. Borosilicate glass upper and lower parts. POM,More Product Information