Ruiskusuodatin, Puradisc™ Aqua 30
Toimittaja: Whatman products (Cytiva)
10462650 10462655
514-0102EA 2030 EUR
514-0102 514-0288
Ruiskusuodatin, Puradisc™ Aqua 30
Suodattimet Ruiskusuodattimet
Puradisc™ Aqua 30 syringe filters are specifically designed for environmental sample filtration prior to dissolved organic carbon (DOC) and chemical oxygen demand (COD) analysis.
  • From the Puradisc FP 30 range
  • Low protein binding, hydrophilic cellulose acetate syringe filter media
  • Prewashed cellulose acetate (CA) syringe filter for reduced organic carbon extractables
  • Designed for sample preparation in support of dissolved organic carbon and chemical oxygen demand analysis
  • Standard inlet and outlet luer connectors
DOC (dissolved organic carbon) and COD (More Product Information