Glass microfibre filters without binder, grade GF/D, Whatman™
Toimittaja: Whatman products (Cytiva)
1823-010 1823-021 1823-024 1823-025 1823-035 1823-047 1823-055 1823-070 1823-090 1823-110 1823-125 1823-142 1823-150 1823-257 1823-915 1823-007 1823-042
512-0427EA 57.6 EUR
512-0427 513-5251 513-5252 513-5235 513-5253 513-5236 513-5255 513-5237 513-5238 513-5239 513-5240 513-0091 516-2421 516-2423 513-5271 512-0666 513-5254
Glass microfibre filters without binder, grade GF/D, Whatman™
Suodattimet Lasikuitusuodattimet
These borosilicate glass filters are fine membrane filters that can also function as a prefilter when used with GF/B.
  • Thick filter media
  • Good loading capacity
  • Fit most filter holders
  • Binder free