Glass microfibre filters without binder, grade GF/B, Whatman™
Toimittaja: Whatman products (Cytiva)
1821-021 1821-024 1821-025 1821-037 1821-042 1821-047 1821-055 1821-070 1821-090 1821-110 1821-125 1821-150 1821-185 1821-915 1821-914 1821-271
513-5262EA 47.9 EUR
513-5262 516-2204 513-5213 513-5214 513-5215 513-5216 513-5217 513-5218 513-5219 513-5220 513-5221 513-5222 516-2222 513-5223 513-0107 512-0665
Glass microfibre filters without binder, grade GF/B, Whatman™
Suodattimet Lasikuitusuodattimet
These borosilicate glass filters are particularly useful for liquid clarification and liquid scintillation counting.
  • Fine particle filter
  • Good loading capacity
  • Binder free