Suodattimet, polyamidisuodatin, SUSP 70
Toimittaja: Ahlstrom-Munksjö
460001 460002 460003
512-6151EA 154 EUR
512-6151 512-6150 512-6149
Suodattimet, polyamidisuodatin, SUSP 70
Suodattimet Lasikuitusuodattimet
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.