Grade 595 ½ qualitative filter papers, Whatman™
Toimittaja: Whatman products (Cytiva)
10311641 10311642 10311643 10311644 10311645 10311647 10311649 10311651 10311652 10311653 10311654 10311656
515-5501EA 48.2 EUR
515-5501 512-3620 512-3621 512-3622 512-3623 512-3624 512-0002 512-3625 512-3626 512-3627 515-5510 515-5511
Grade 595 ½ qualitative filter papers, Whatman™
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
A prepleated filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples, e.g., for ICP/AAS analysis).
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is