Vakuumisuodatusyksiköt
Toimittaja: Sartorius
16510 16511
512-3310EA 182 EUR
512-3310 512-3311
Vakuumisuodatusyksiköt
Suppilot Membraanisuodatinsuppilot
PC, voidaan autoklavoida.
Alle 200 ml vesiliuosten kirkastamiseen tai steriilisuodatukseen.
512-3310 is complete with receiver flask of ±250 ml for sub-pressure as well as with slight over-pressure.
512-3311 without receiver, can be used in combination with Combisart manifolds.
For sterile filtrations- the filter holder includes a glass fibre filter that enables sterile venting, thus avoiding contamination of the sterile filtrate.