Buchner funnels
Toimittaja: DIXON SCIENTIFIC
232040112 232040113 232040122 232040123 232040133
511-0801EA 106 EUR
511-0801 511-0802 511-0805 511-0806 511-0810
Buchner funnels
Suppilot Suodatussuppilot Büchner-suppilot
Borosilicate glass
  • With sintered glass disc
  • Very good chemical resistance
  • High temperature resistance
For paper or membrane filtration disc.