Filtration funnels, Büchner type
Toimittaja: SciLabware
3722/06M 3722/08M 3722/02M 3720/02M 3720/04M 3720/06M 3620/02 3620/04 3620/06 3620/08 3620/10 3630/01 3630/02 3630/04 3630/06 3630/08 3630/10 3700/01 3700/02 3700/04 3700/06 3700/08 3700/10 3710/01 3710/02 3710/04 3710/06 3720/08M 3722/04M
511-0783EA 805 EUR
511-0783 511-0784 511-0781 511-0778 511-0779 511-0780 BIBB3620/02 BIBB3620/04 BIBB3620/06 BIBB3620/08 BIBB3620/10 BIBB3630/01 BIBB3630/02 BIBB3630/04 BIBB3630/06 BIBB3630/08 BIBB3630/10 BIBB3700/01 BIBB3700/02 BIBB3700/04 BIBB3700/06 BIBB3700/08 BIBB3700/10 BIBB3710/01 BIBB3710/02 BIBB3710/04 511-0246 BIBB3720/08M 511-0039
Filtration funnels, Büchner type
Suppilot Suodatussuppilot Büchner-suppilot
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
For convenient filtration of solutions.