VWR®, Filtering Flasks, with Glass Hose Connection
Toimittaja: VWR Collection
511-0260EA 26.4 EUR
511-0260 511-0261 511-0262 511-0263 511-0264
VWR®, Filtering Flasks, with Glass Hose Connection
Kolvit ja erlenmeyerit Imupullot
Borosilicate 3.3 glass.
  • Suitable for hot and cold filtration
  • Highly resistant to chemicals and thermal shock
  • High temperature resistance
  • White labelling area
Due to the heavy wall thickness the flask is vacuum tight.
Sertifikaatit: ISO 6556.