Filter candles, conical, VitraPOR®
Toimittaja: ROBU GLASFILTER
18201 18202 18203 18204 18311 18312 18313 18314 18324 18212 18213 18214 18300 18301 18302 18303 18304 18310 18320 18321 18210 18323 18200 18322 18211
511-0201EA 107 EUR
511-0201 511-0202 511-0203 511-0204 511-0211 511-0212 511-0213 511-0214 ROBU18324 ROBU18212 ROBU18213 ROBU18214 ROBU18300 ROBU18301 ROBU18302 ROBU18303 ROBU18304 ROBU18310 ROBU18320 ROBU18321 ROBU18210 ROBU18323 ROBU18200 ROBU18322 ROBU18211
Filter candles, conical, VitraPOR®
Suodattimet Suodatinputket
These conical filter candles are available with or without tube.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of borosilicate glass 3.3.