ROBU® VitraPOR® Glass Filter Discs, Standard
Toimittaja: ROBU GLASFILTER
15100 15101 15102 15103 15104 15050 15200 15201 15202 15203 15204 15250 15251 15252 15253 15254 15300 15301 15302 15303 15304 15400 15401 15402 15403 15404 15500 15501 15502 15503 15504 15600 15601 15602 15603 15604 15653 15654 15700 15701 15702 15703 15704 15800 15801 15802 15803 15804 15900 15901 15902 15903 15904 15120 15121 15122 15123 15124 15655 15659 15705 15709 15805 15809 15905 15909 15651 15650 15129 15052 15053 15054 15055 15105 15109 15110 15111 15112 15113 15114 15115 15652 15125 15205 15209 15255 15259 15305 15309 15405 15409 15505 15509 15605 15609 15119 15051 52503 52704 52703 52702 52701 52700 52604 52603 52602 52601 52801 52504 52802 52502 52501 52500 52404 52403 52402 52401 52400 52304 52303 52302 52600 53004 53804 53803 53802 53801 53800 53504 53503 53502 52800 53500 53003 53002 53001 53000 52904 52903 52902 52901 52900 52804 52803 53501 51501 51703 51702 51701 51700 51604 51603 51602 51601 51600 51504 52300 51502 51801 51500 51404 51403 51402 51401 51400 51304 51303 51302 51301 51503 52002 52203 52202 52201 52200 52104 52103 52102 52101 52100 51704 52003 51800 52001 52000 51904 51903 51902 51901 51900 51804 51803 51802 52004 51300 52301 52204 1505F 1510F 1511F 1512F 1520F 1525F 1530F 5150F 5160F 5170F 5180F 5190F 5200F 1540F 1550F 1560F 1565F 1570F 1580F 1590F 5130F 5140F
511-0010EA 5.2 EUR
511-0010 511-0011 511-0012 511-0013 511-0014 511-0015 511-0020 511-0021 511-0022 511-0023 511-0024 511-0030 511-0031 511-0032 511-0033 511-0034 511-0040 511-0041 511-0042 511-0043 511-0044 511-0050 511-0051 511-0052 511-0053 511-0054 511-0060 511-0061 511-0062 511-0063 511-0064 511-0070 511-0071 511-0072 511-0073 511-0074 511-0078 511-0079 511-0080 511-0081 511-0082 511-0083 511-0084 511-0090 511-0091 511-0092 511-0093 511-0094 511-0100 511-0101 511-0102 511-0103 511-0104 511-0110 511-0111 511-0112 511-0113 511-0114 ROBU15655 511-0247 ROBU15705 ROBU15709 511-0345 511-0346 511-0347 511-0348 ROBU15651 ROBU15650 511-0344 ROBU15052 ROBU15053 ROBU15054 ROBU15055 ROBU15105 ROBU15109 511-0306 511-0307 511-0308 511-0309 511-0310 511-0341 ROBU15652 511-0343 ROBU15205 ROBU15209 ROBU15255 ROBU15259 ROBU15305 ROBU15309 ROBU15405 ROBU15409 ROBU15505 ROBU15509 ROBU15605 ROBU15609 511-0342 ROBU15051 511-0461 511-0472 511-0471 511-0470 511-0469 511-0468 511-0467 511-0466 511-0465 511-0464 511-0474 511-0462 511-0475 511-0460 511-0459 511-0458 511-0457 511-0456 511-0455 511-0454 511-0453 511-0452 511-0451 511-0450 511-0463 511-0487 511-0497 511-0496 511-0495 511-0494 511-0493 511-0492 511-0491 511-0490 511-0473 511-0488 511-0486 511-0485 511-0484 511-0483 511-0482 511-0481 511-0480 511-0479 511-0478 511-0477 511-0476 511-0489 511-0397 511-0417 511-0416 511-0415 511-0410 511-0409 511-0408 511-0407 511-0406 511-0405 511-0400 511-0448 511-0398 511-0420 511-0396 511-0395 511-0394 511-0393 511-0392 511-0391 511-0390 511-0389 511-0388 511-0387 511-0399 511-0435 511-0446 511-0445 511-0444 511-0443 511-0442 511-0441 511-0440 511-0439 511-0438 511-0418 511-0436 511-0419 511-0434 511-0433 511-0432 511-0431 511-0430 511-0429 511-0428 511-0427 511-0426 511-0425 511-0437 511-0386 511-0449 511-0447 ROBU1505F ROBU1510F ROBU1511F ROBU1512F ROBU1520F ROBU1525F ROBU1530F ROBU5150F ROBU5160F ROBU5170F ROBU5180F ROBU5190F ROBU5200F ROBU1540F ROBU1550F ROBU1560F ROBU1565F ROBU1570F ROBU1580F ROBU1590F ROBU5130F ROBU5140F
ROBU® VitraPOR® Glass Filter Discs, Standard
Suodattimet Suodatinkiekot
VitraPOR® glass filters are precision-engineered porous glass components for effective filtration in laboratory and industrial settings.
  • Made from high-purity borosilicate glass
  • Available in different porosities
  • Chemically resistant and autoclavable
  • Reusable and environmentally friendly
  • Custom sizes and designs on request
  • Excellent flow rates and mechanical strength
ROBU VitraPOR glass filters made of borosilicate glass offer outstanding chemical resistance, thermal stability, and precise filtration performance. Manufactured with a uniform pore
More Product Information
Order Now
TEKNISET TIEDOT
Hakemus Kaasujakauma nesteissä pienellä paineella, karkeasuodatus Karkeasuodatus, kaasujakauma, nestejakauma, kaasupuhdistus Alustava hienosuodatus, elohopeasuodatus, kiteisten kristallituotteiden valmistelu Keskikarkeiden sakkojen analysointi, kaasun hienosuodatus, analyyttinen suodatus, erikoissuodatus Analyyttinen hienosuodatus, hienojakoisten sakkojen analyysit ja valmistelu, sulku- ja paluuventtiili elohopealle Gas distribution in liquids at low pressure, coarse filtration Preparative fine filtration, mercury filtration, preparative work with coarse crystalline products Analytical work with medium fine precipitates, fine gas filtration, analytical filtration, special filtration Analytical fine filtration, analytical and preparative work with the finest precipitates, return and stop valve for mercury Bacteria filtration, sterile filtration Liquid and gas distribution, substrates for solids in fluid systems Coarse filtration, gas distribution, liquid distribution, gas cleaning Fine filtration
Nimellinen huokoskoko 160…250 µm 100…160 µm 40…100 µm 16…40 µm 10…16 µm 160…250 µm 40…100 µm 16…40 µm 10…16 µm 1…1,6 µm 250…500 µm 100…160 µm 4 - 5,5 µm
Environmentally Preferable
Huokoisuus 0 1 2 3 4 0 2 3 4 5 00 1 Fine