VWR®, Testiseulat, 305×60 mm
Toimittaja: VWR Collection
510-2837EA 266 EUR
510-2837 510-2838 510-2839 510-2840 510-2841 510-2842 510-2843 510-2844 510-2845 510-2846 510-2847 510-2848 510-2849 510-2850 510-2851 510-2852 510-2853 510-2854 510-2855 510-2856 510-2857 510-2858 510-2859 510-2860 510-2861 510-2862 510-2863 510-2864 510-2865 510-2866 510-2867 510-2868 510-2869 510-2870 510-2871 510-2872 510-2873 510-2874 510-2875 510-2876 510-2877 510-2878 510-2879 510-2880 510-2881 510-2882 510-2883 510-2884 510-2885 510-2886 510-2887 510-2888 510-2889 510-2890 510-2891 510-2892 510-2893 510-2894 510-2895 510-2896 510-2897 510-2898 510-2899 510-2900 510-2901 510-2902 510-2903 510-2904 510-2905 510-2906 510-2907 510-2908 510-2909 510-2910 510-2911 510-2912 510-2913 510-2914 510-2915 510-2916 510-2917 510-2918 510-2919 510-2920 510-2921 510-2922 510-2923 510-2924 510-2925 510-2926 510-2927 510-2928
VWR®, Testiseulat, 305×60 mm
Seulat Testisiivilät
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Toimitustiedot: More Product Information