VWR®, Test Sieves, 300×30 mm, with Square Holes
Toimittaja: VWR Collection
510-2649EA 281 EUR
510-2649 510-2650 510-2651 510-2652 510-2653 510-2654 510-2655 510-2656 510-2657 510-2658 510-2659 510-2660 510-2661 510-2662 510-2663 510-2664 510-2665 510-2666 510-2667 510-2668 510-2669 510-2670 510-2671 510-2672 510-2673 510-2674 510-2675 510-2676 510-2677 510-2678 510-2679 510-2680 510-2681 510-2682 510-2683 510-2684 510-2685 510-2686 510-2687 510-2688 510-2689
VWR®, Test Sieves, 300×30 mm, with Square Holes
Seulat Testisiivilät
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.