VWR®, Test Sieves, 150×50 mm
Toimittaja: VWR Collection
510-0578EA 182 EUR
510-0578 510-0579 510-0580 510-0581 510-0582 510-0583 510-0584 510-0585 510-0586 510-0587 510-0588 510-0589 510-0590 510-0591 510-0592 510-0593 510-0594 510-0595 510-0596 510-0597 510-0598 510-0599 510-0600 510-0601 510-0602 510-0603 510-0604 510-0605 510-0606 510-0607 510-0554 510-0555 510-0556 510-0557 510-0558 510-0559 510-0560 510-0561 510-0562 510-0563 510-0564 510-0565 510-0566 510-0567 510-0568 510-0569 510-0570 510-0571 510-0572 510-0573 510-0574 510-0575 510-0576 510-0577
VWR®, Test Sieves, 150×50 mm
Seulat Testisiivilät
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Toimitustiedot: More Product Information