Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104]
Toimittaja: Tonbo Biosciences
50-0454-U100 50-0454-U025
50-0454-U100EA 127 EUR
50-0454-U100 50-0454-U025
Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104]
Vasta-aineet
The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. 
The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to beMore Product Information