Anti-CD8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SK1]
Toimittaja: Tonbo Biosciences
50-0087-T025 50-0087-T100 50-0087-T500
50-0087-T025EA 127 EUR
50-0087-T025 50-0087-T100 50-0087-T500
Anti-CD8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SK1]
Vasta-aineet
The SK1 antibody is specific for the 32-34 kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information