Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Hit3a]
Toimittaja: Tonbo Biosciences
50-0039-T100 50-0039-T025
50-0039-T100EA 63.9 EUR
50-0039-T100 50-0039-T025
Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Hit3a]
The Hit3a antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing cell proliferation, differentiation, production ofMore Product Information