N,N-Dimetyyliformamidi ≥99.9%, LiChrosolv® for liquid chromatography, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ100

5.43897.1000 5.43897.2500
5.43897.1000EA 161 EUR
5.43897.1000 5.43897.2500
N,N-Dimetyyliformamidi ≥99.9%, LiChrosolv® for liquid chromatography, Supelco®
N,N-Dimetyyliformamidi
Kaava: HCON(CH₃)₂
Kiehumispiste: 153 °C (1013 hPa)
Sulamispiste: –61 °C
MDL numero: MFCD00003284
CAS-numero: 68-12-2
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity (GC) ≥ 99.9 %
Water ≤ 0.03 %
Transmission (at 275 nm) ≥ 65 %
Transmission (at 300 nm) ≥ 90 %
Transmission (at 350 nm) ≥ 99.5 %
Transmission (at 400 nm) ≥ 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR