J.T.Baker®, Robotic Tips Type TE
Toimittaja: Avantor
49002-0000EA 2000 EUR
49002-0000 49002-0006 49002-0007 49002-0106 49003-0000 49003-0006 49003-0007 49003-0106 49003-0107 49006-0000 49006-0006 49006-0106 49006-0107 49033-0000 49033-0006 49033-0106 49102-0000 49103-0000 49002-1106 49002-1107 49003-1106 49003-1107 49006-1106 49006-1107 49033-1106 49102-1106 49103-1106 49106-1106 49102-0006 49103-0006 49106-0000 49133-0000 49002-3106 49003-3106 49102-3106 49103-3106 49002-2006 49002-2106 49003-2106 49024-0008 49002-0117 49026-0017 49026-0117
J.T.Baker®, Robotic Tips Type TE
Pipetinkärjet
Suitable for use on Tecan Freedom EVO or Tecan Fluent. Available in conductive and non conductive versions, with or without filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Ideal for high throughput analysis
  • Packed in cardboard or blister
J.T.Baker® robotic tips type TE are suitable for use on Tecan Freedom EVO (typically uses tips in S-Tray = hanging tray), Tecan Fluent (typically uses tips in C-Tray for ANSI/SLAS format tip box) andMore Product Information