ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Millipore®

Toimittaja: Merck Millipore Calbiochem

481973-1 481973-100 481973-25 481973-50 481973-500
481973-1EA 1980 EUR
481973-1 481973-100 481973-25 481973-50 481973-500
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Millipore®
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Kaava: C₂₁H₂₆N₇Na₄O₁₇P₃
MDL numero: MFCD00036263
CAS-numero: 2646-71-1
EINECS: 220-163-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR