Autoclaves, front loading, D-Line
Toimittaja: TUTTNAUER
2840EL-D 3850EL-D 2840ELC-G-D 3870ELC-G-D 5075ELC-G-D 5075EL-D 3870EL-D
481-0555EA 10140 EUR
481-0555 481-0567 481-0849 TUTT3870ELC-G-D TUTT5075ELC-G-D TUTT5075EL-D TUTT3870EL-D
Autoclaves, front loading, D-Line
Sterilisaattorit Autoklaavit (höyrysterilointilaitteet)
Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceutical, food, medical and biotechnological industries. This advanced laboratory line is a quality range of reliable bench top (front loading) and vertical (top loading) models with chamber volumes from 28 to 160 litres that provides a single solution for the full spectrum of sterilisation needs. The autoclaves can be fully configured from general use to customer-specific applications. Units from either the Essential or Hi End ranges can beMore Product Information