VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Toimittaja: VWR Collection
479-7019EA 5.2 EUR
479-7019 479-7020 479-7021 479-7022 479-7023 479-7024 479-7025 479-7026 479-7027 479-7028 479-7029 479-7030 479-7031 479-7032 479-7033 479-7034 479-7035
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Laatikot Kryogeenipullolaatikot
Cardboard, coated.
  • Waterproof coating
  • Freezable to –140 °C
  • Available with or without dividers
  • In different heights and five colours
For storage of cryovials at low temperatures.