VWR® Cryoboxes, Double Size, 133x257 mm
Toimittaja: VWR Collection
479-7006EA 8.4 EUR
479-7006 479-7007 479-7008 479-7009 479-7010 479-7011 479-7012 479-7013 479-7014 479-7015 479-7016 479-7017 479-7018
VWR® Cryoboxes, Double Size, 133x257 mm
Laatikot Kryogeenipullolaatikot
Corrugated cardboard box.
  • Waterproof coating
  • Freezable to –140 °C
  • Available with and without dividers
  • In different heights and five colours
For storage of cryovials at low temperatures.