High-speed barcode reader, VisionMate™ HSX
Toimittaja: THERMO MATRIX TECHNOLOGIES
312850
479-2448EA 18800 EUR
479-2448
High-speed barcode reader, VisionMate™ HSX
Viivakoodijärjestelmät
The Thermo Scientific™ VisionMate™ HSX High Speed Barcode Reader decodes 2D tube barcodes in as little as one second. Compatible with Matrix™ and Nunc™ tubes in 24, 48, 96, and 384 format racks.
  • Pre-loaded rack configurations allow you to rapidly read Matrix™ and Nunc™ racks of tubes without additional set up steps
  • Can decode both 2D and linear rack barcodes, providing additional tracking options
  • Customizable LED visual feedback provides the user with instant notification on successful reads, as well as notification of any read errors
  • Included magnetic
More Product Information
Order Now
TEKNISET TIEDOT
Environmentally Preferable Low Manufacturing Impact
Liitännät USB 2.0+
Muisti No internal storage capacity; data is stored on the user's computer
Virta USB Powered
L×S×K 200×150×160 mm
Ympäristösertifikaatti NAR Certificate of Retirement