VWR®, Cryoboxes and dividers
Toimittaja: VWR Collection
478-0255EA 26.4 EUR
478-0255 478-0244 479-2216 479-2217 479-2219 479-2220 479-2221 479-2223 479-2224 479-2225 479-2227 479-2228 479-2213 479-2215
VWR®, Cryoboxes and dividers
Laatikot Kryogeenipullolaatikot
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.