CryoELITE® Cryogenic Vials with Pre-Inserted Barcodes, Freestanding, External Thread
Toimittaja: DWK Life Sciences
W985863-BC W985864-BC W985865-BC W985866-BC W985867-BC W985868-BC
479-2081EA 1590 EUR
479-2081 479-2082 479-2083 479-2084 479-2085 479-2086
CryoELITE® Cryogenic Vials with Pre-Inserted Barcodes, Freestanding, External Thread
Näytevialit ja pienet näyteastiat
Wheaton CryoELITE® Cryogenic Freestanding Vials have a Loctagon™ Vial Skirt that provides stability in the freestanding position and locks into the CryoELITE® Benchmate Rack in order to provide easy open and close with one hand.
The 2D Data Matrix Bar Code Bottom Inserts allows for immediate bar coding of samples. The 2D Data Matrix Bar Code Insert is locked into place in the bottom of the vial. The insert is made of the same polypropylene as the vials themselves and is laser-etched for durability.
These vials are supplied with Wheaton 2D Data Matrix Bar Codes inserted. The Bar CodesMore Product Information