Kryoputket Cryobank ja Bank-It™, Nunc™
Toimittaja: Thermo Fisher Scientific
374086 374074 374088 374089 374078 374080 374081 374110 374120 374130 374025 374026 374027 374220 374221 374222 374223 374258 374260 374261 374259 374220LOCK 374221LOCK 374115 374221-BR 374220-BR
479-0588EA 870 EUR
479-0588 479-0590 479-0592 479-0593 479-0594 479-1081 479-1082 479-1083 479-1084 479-1085 479-1077 479-1078 479-1079 479-1086 479-1087 479-1088 479-1089 479-1328 479-1330 479-1331 479-1329 479-2071 479-2072 479-2070 479-2495 479-2288
Kryoputket Cryobank ja Bank-It™, Nunc™
Näytevialit ja pienet näyteastiat
These cryogenic vials have the same features as the standard cryotubes. They are designed for long term low temperature storage. Some vials have laser etched unique 2D barcodes that allow for tracking of the samples. The codes are permanently attached so they do not separate from the vials. Barcoded vials feature standard microplate format racks.
  • Vials can be labelled with a cryopen
  • Microplate format is compatible with your existing multichannel manual pipettes and automated liquid handling equipment
  • Low binding surface allows maximum recovery of low