Kryolaatikko, pakastinlaatikko, CryoFile™, WHEATON®
Toimittaja: DWK Life Sciences
W651600 W651600-XL W651601 W651601-XL W651602 W651602-XL W651603 W651603-XL W651604 W651604-XL W651605 W651605-XL
479-2000EA 281 EUR
479-2000 479-2001 479-2002 479-2003 479-2004 479-2005 479-2006 479-2007 479-2008 479-2009 479-2010 479-2011
Kryolaatikko, pakastinlaatikko, CryoFile™, WHEATON®
Laatikot Kryogeenipullolaatikot
High quality chip-board, spray coated with polyethylene.
  • Water repellent material allows for longer durability
  • Numbering system printed on lid and bottom of the box
  • Withstands cryogenic temperatures to –196 °C
  • Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.