Corning® CoolCell® containers
Toimittaja: Corning
432000 432001 432002 432003 432004 432005 432006 432007 432008 432009 432010 432011 432138
479-1840EA 155.6 EUR
479-1840 479-1841 479-1842 479-1843 479-1844 479-1845 479-1848 479-1849 479-1850 479-1851 479-1918 479-1919 CORN432138
Corning® CoolCell® containers
Laatikot Kryogeenipullolaatikot
CoolCell® alcohol-free cell freezing containers ensure standardised controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.
  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 °C freezer
  • Quick re-use time, ready to use again after five minutes
Proven for use with a varietyMore Product Information