VWR®, Cryoboxes, without or with Dividers
Toimittaja: VWR Collection
479-1382EA 363 EUR
479-1382 479-1383 479-1384 479-1385 479-1386
VWR®, Cryoboxes, without or with Dividers
Laatikot Kryogeenipullolaatikot
Made of white cardboard.
  • Inexpensive to buy, inexpensive to ship, space saving in the laboratory
  • Available in the most sold size with a height of 51 mm
  • Square, size of bottom 129×129 mm, size of top 134×134 mm
  • Fits into all storage rack systems
Consists of a lid and a base which are delivered flat. Simply by following the instructions on the lid, 1-2-3 and you have a stable, high quality cryobox.