IsoTherm-System® Sample Cooling System
Toimittaja: EPPENDORF
3880001.018 3880001.166 3880001.174
479-1120EA 195 EUR
479-1120 479-1152 479-0003
IsoTherm-System® Sample Cooling System
Laatikot Pöytämalliset jäähdyttimet
Cool samples safely and efficiently using the Eppendorf® IsoTherm-System. Choose between IsoPacks for 0 °C and –21 °C and use the IsoRack for centrifuging, transporting, or storing, ensuring constant temperature maintenance throughout your workflow.
  • Efficiently cool samples at a constant temperature and then thaw them gently
  • IsoRack suitable for centrifuging in a plate rotor with IsoRack adapter
  • IsoRack serves as a work rack when filling micro test tubes and for provisioning, sorting, distributing, transporting, storing, or freezing samples in micro test