Cryogenic vessel, Bio-Cane®
Toimittaja: Thermo Fisher Scientific
CK509X3 CK509X4 CK509X2
478-0804EA 2620 EUR
478-0804 478-0805 478-0803
Cryogenic vessel, Bio-Cane®
Nestetyppisäiliöt
Cryogenic vessels are ideal to store cells, tissues and biologicals in an economical and biological manner.
  • Six stainless-steel canisters to accommodate vials
  • Canisters are numbered and colour-coded to simplify sample identification
  • Durable aluminium construction and vacuum insulation
  • Secure locking clasp and narrow-mouth design minimises LN₂ evaporation