MES ≥99% (by titrimetric analysis on dry substance), ULTROL®, Millipore®

Toimittaja: Merck Millipore Calbiochem

475893-1 475893-100
475893-1EA 1020 EUR
475893-1 475893-100
MES ≥99% (by titrimetric analysis on dry substance), ULTROL®, Millipore®
MES
Kaava: C₆H₁₃NO₄S
Sulamispiste: 316 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006181
CAS-numero: 4432-31-9
EINECS: 224-632-3
Merck Index: 13,05929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR