Lenvatinib mesylate 98%

Toimittaja: Thermo Fisher Scientific

474520010 474522500
474520010.EA 473 EUR
474520010. 474522500.
Lenvatinib mesylate 98%
Lenvatinib mesylate
Kaava: C₂₂H₂₃ClN₄O₇S
Storage Temperature: Jääkaappi
MDL numero: MFCD18633219
CAS-numero: 857890-39-2

Order Now

Testitulosten erittely

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR