Platina nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support

Toimittaja: Thermo Fisher Scientific

47371.03 47371.06
47371.03EA 589 EUR
47371.03 47371.06
Platina nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support
Platina
Kaava: Pt
MDL numero: MFCD00011179
CAS-numero: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR